Learn More
Enzo Life Sciences DPN (50mg). CAS: 1428-67-7
SDP
Supplier:  Enzo Life Sciences BMLGR2480050
Encompass_Preferred
Selective estrogen receptor ERβ agonist displaying 70-fold greater selectivity over ERα(EC50 = 0.85 and 66nM, respectively). It displays neuroprotective activity in various in vitro and in vivo models. Alternative name: Diarylpropionitrile, 2,3-bis(4-Hydroxyphenyl)-propionitrile. Purity: ≥98% (TLC). Solubility: Soluble in DMSO (10mg/ml) or 100% ethanol (1.5mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-0953