Learn More
Abcam DPN, ERbeta agonist, 50MG
Supplier:  Abcam AB12016650MG
Encompass_Preferred
MW 239.27 Da, Purity >99%. Potent, selective ERβ agonist (EC₅₀ values are 0.85 and 66 nM for ERβ and ERα, respectively). Cardioprotective and neuroprotective in vivo.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7827