Learn More
ABclonal Technology DPPA2 Rabbit pAb
SDP
Supplier:  ABclonal Technology A14966500UL
Encompass_Preferred
Predicted to enable chromatin binding activity. Predicted to be involved in system development. Predicted to act upstream of or within several processes, including lung-associated mesenchyme development, positive regulation of stem cell proliferation, and regulation of histone methylation. Located in nucleoplasm.
Catalog No. 50-001-08605