Learn More
Echelon Biosciences Research Labs DPPC 1 g
SDP
Supplier:  Echelon Biosciences Research Labs L11161G
Encompass
1,2-Dipalmitoyl-sn-glycero-3-phosphocholine (DPPC) is a phosphocholine with two palmitic acids on its head-group and is used in formulation of medicines.Phosphatidylcholine (PC) is generally the most abundant lipid in animal cell membranes providing structural framework. PC is more common in the outer leaflet where it functions as part of the permeability barrier. PC is also the primary substrate of Phospholipase D enzymes which produce the signaling lipids phosphatidic acid and lysophosphatidic acid. This item is non returnable
Catalog No. 50-720-489