Learn More
Kyfora Bio DPTAP 500 MG
Supplier:  Kyfora Bio 14487500
Encompass_Preferred
DPTAP Chloride (1,2-dipalmitoyl-3-trimethylammonium-propane Chloride) is a cationic lipid and an analogue of DOTAP. It can be used in the formation of liposomes and microbubbles that encapsulate nucleic acids, such as miRNA, as an efficient gene delivery system.Key application(s): Gene delivery, miRNA delivery
Catalog No. 50-255-9835