Learn More
ACROBiosystems Recombinant Protein;Cynomolgus DR3 / TNFRSF25 Protein, Fc Tag;HEK293;1MG;TN5,TNFRSF25,DR3,APO3,DDR3,TNFRSF12,WSL,WSL1,TNFRSF-25,DR-3,APO-3,DDR-3,TNFRSF-12,WSL-1
Supplier:  ACROBiosystems TN5C52571MG
Encompass_Preferred
Recombinant Protein;Cynomolgus DR3 / TNFRSF25 Protein, Fc Tag;1MG;Cynomolgus DR3, Fc Tag (TN5-C5257) is expressed from human 293 cells (HEK293). It contains AA Gln 20 - Gly 203 (Accession # G7NTB5).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 46.3 kDa. The protein migrates as 50 kDa under reducing (R) condition (SDS-PAGE).
Catalog No. 50-165-9112