Learn More
ABclonal Technology DR4 Rabbit mAb
SDP
Supplier:  ABclonal Technology A2470320UL
Encompass_Preferred
The protein encoded by this gene is a member of the TNF-receptor superfamily This receptor is activated by tumor necrosis factor-related apoptosis inducing ligand (TNFSF10 TRAIL) and thus transduces cell death signal and induces cell apoptosis Studies with FADD-deficient mice suggested that FADD a death domain containing adaptor protein is required for the apoptosis mediated by this protein
Catalog No. 50-000-09670