Learn More
Med Vet International Respiratory Test Kit QuickVue Influenza A + B, CLIA Waived, 20183, 25 Tests/Kit
Supplier:  Med Vet International 489402KT
Encompass_Preferred
Results within 10 minutes Provides differential results for type A and type B viral antigens The QuickVue Influenza A+B test has been shown to detect cultured avian influenza viruses, including avian influenza A subtype H5N1 virus Two-color result Kit includes: 25 individually packaged test strips; 25 reagent solution vial; 25 reagent tubes; 25 disposable pipettes; 25 sterile nasal swabs; positive Type A control swab; positive Type B control swab; negative control swab; package insert; and procedure card Room temperature storage
Catalog No. 50-304-4348
 Restricted Item