Learn More
Med Vet International Surgical Drape 3M Steri-Drape Small Drape With Aperture 22 W X 25 L inch Sterile, 1092, Each
Supplier:  Med Vet International 5727EA
Encompass_Preferred
The sterile surgical drapes with polyethylene film create an impenetrable barrier that resists fluid leaks They minimize the risk of infection by reducing fluid exposure to the skin The aperture with adhesive conforms to the body producing a reliable hold They prevent patient infection at the wound site and cross-contamination The disposable drape sheets have a matte finish to reduce glare
Catalog No. 50-282-9648