Learn More
Med Vet International Sontara Drape, 55" X 77", Non-Sterile, J1060C
Supplier:  Med Vet International JOR1060C
Encompass_Preferred
Sontara Drape is a popular DuPont product with a soft, cloth-like feel. It is fluid repellent, but fluids will strike through with extended exposure. It can be readily autoclaved. Sontara can also be coated with a clear poly film to make it impervious. However, the impervious material cannot be autoclaved.
Catalog No. 50-283-1819