Learn More
New England Biolabs, Inc. DrdI – 300 units
SDP
Supplier:  New England Biolabs, Inc. R0530S
Encompass
A restriction endonuclease that recognizes the sequence GACNN_NN^NNGTC.
Catalog No. 50-812-192