Learn More
Cayman Chemical DREADD AgonIst 21 10mg
SDP
Supplier:  Cayman Chemical 1890710 MG
Encompass_Preferred
Selectively activates hM3Dq (EC50 = 1.7 nM), a DREADD derived from the human muscarinic acetylcholine M3 receptor
Catalog No. 50-298-3930