Learn More
Med Vet International IV Specialty Dressing 3M Tegaderm Film 2 X 2-1/4 inch Sterile, 1610, 100/Box
Supplier:  Med Vet International 452711BX
Encompass_Preferred
Fun Tegabear design helps ease child patient's anxieties Transparent film over insertion site allows easy skin and site monitoring Sterile microporous documentation label supports good catheter care practice Two sterile soft cloth tape strips help to anchor catheter and infusion lines Waterproof to protect against external contamination Deep notch design seals around catheter to protect puncture site Design prevents tenting of dressing and stick-to-itself wastage
Catalog No. 50-283-1316