Learn More
Med Vet International Orthopedic Padding Adhesive 3M Reston 7.875 X 11.75 inch Foam Nonsterile, 1561H, 5/Pack
Supplier:  Med Vet International 5789PK
Encompass_Preferred
High support Adhesive Self-adhering foam designed to distribute body weight to help prevent skin breakdown, pressure ulcers and to prevent skin damage from splints, casts and prostheses Can also be used as a padding for beds, chairs and operating room tables High support pad can be cut, layered and shaped to be adapted for many applications Reston self-adhering foam products should not be placed in direct contact with an open wound
Catalog No. 50-283-1487
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.