Learn More
Med Vet International Dynarex Sterile Curafoam Ag Silver Foam Dressing, 4" X 4", 100/Case
Supplier:  Med Vet International 3047
Encompass_Preferred
A sterile, highly absorbent, non-bordered dressing that provides a warm, moist wound environment with continuous antimicrobial protection to prevent colonization on the dressing. Non-adherent foam with an ionic silver compound to allow exudate to be absorbed while providing an antimicrobial barrier. Maintains a warm, moist wound environment with continuous antimicrobial protection to prevent colonization. Highly absorbent foam for management of chronic and acute wounds at stages 2, 3 and 4. Primary dressing for wounds at risk of infection or re-infection. Ideal for the management of pressure ulcers, surgical wounds, and first and second degree burns.
Catalog No. 50-283-1387