Learn More
Med Vet International Transparent Film Dressing 3M Tegaderm 6 X 8 inch Frame Style DelIVery Rectangle Sterile, 1628, Each
Supplier:  Med Vet International 121653EA
Encompass_Preferred
3M™ Tegaderm™ film dressing has a frame style delivery system for better control and accurate positioning It forms a protective sterile barrier to lock out external contaminants for healing to occur Transparent construction provides high visibility for easy observation of the affected site The adhesive is latex-free and hypoallergenic to accommodate different skin types It bonds well to hold medical devices like catheters firmly in place It peels off gently for reduced pain in between dressing changes Wear time is up to 7 days
Catalog No. 50-282-9962