Learn More
Med Vet International Curad Xeroform Dressing, 5in x 9in, Each
Supplier:  Med Vet International CUR253590H
Encompass_Preferred
Curad Xeroform is a U.S.P absorbent fine mesh petrolatum gauze that is impregnated wtih 3% Bismuth Tribromophenate deoderizing agent that helps maintain moisture. This gauze dressing clings, conforms, and allows minimal drainage to pass through to secondary dressings. Curad Xeroform is a non-adherent dressing designed to help prevent a secondary dressing from "sticking" to wound/periwound skin and helps prevent trauma from dressing changes. Xeroform is ideal for non-draining to lightly draining wounds including lacerations, minor or partial thickness burns, abrasions, skin graft sites, and suture wounds. Size: 5x9 Quantity: 1
Catalog No. 50-209-1860