Learn More
Med Vet International Dri-I Schirmer Test, J1192B, 100/Pack
Supplier:  Med Vet International JOR1192B
Encompass_Preferred
Now with blue absorbent indicator strip. - Indicated at 5mm, the blue absorbent ink moves up with the tear film level giving a clearer indication of tear film production.
Catalog No. 50-005-81227