Learn More
Cayman Chemical DRIbulose5phosphatesodIu 10mg
SDP
Supplier:  Cayman Chemical 2142310 MG
Encompass_Preferred
An intermediate in the pentose phosphate pathway, carbon fixation in photosynthetic organisms, and the biosynthesis of lipopolysaccharide, amino acids, secondary metabolites, and antibiotics
Catalog No. 50-299-4072