Learn More
Accuflow Systems 29 PC STEPPED TIP DRILL BIT SET
Supplier:  Accuflow Systems 63200
Encompass_Preferred
New from Steel Vision Tools comes these new groundbreaking drill bit sets. One of the very first to hit the market of its kind is the stepped tip drill bit design. Each drill bit has a small step bit tip. Each fluted step tip is in proportion to the bit size. Even on round, curved or uneven surfaces the stepped cutter bits will glide through metal without a pilot hole and drastically reduce walking. The Tungsten Cobalt hi-moly alloy allows for easier penetration and precise cuts. Save time, space and money with these new stepped cutter drill bit sets from SVT. -Outlasts split point bits 4-1- Never needs pilot hole- Mechanics length makes them less prone to breakage- M35 Tungsten-Cobalt hi-moly alloyIncludes all of these sizes63201 1/16″ Stepped Tip Drill Bit63202 5/64″ Stepped Tip Drill Bit63203 3/32″ Stepped Tip Drill Bit63204 7/64″ Stepped Tip Drill Bit
Catalog No. 50-223-6806