Learn More
Grainger Drill Mill 12.00mm Carbide Bright
Supplier:  Grainger 16T656
Encompass_Preferred
The Micro 100 Drill Mill Carbide is a high-performance end mill for milling and drilling. It has a TiAlN coating for tool life and heat resistance. With a 1/8" diameter, this 2-flute end mill has a straight shank and is center cutting. Made in the USA, it has a 2-1/2" overall length and can machine steel, stainless steel, cast iron, aluminum, copper, brass, bronze, plastic, and graphite. Its universal shank fits CNC machines, manual milling machines, and drill presses. The carbide construction ensures wear resistance and strength for demanding applications.
Catalog No. 50-276-6699