Learn More
Grainger Drill Mill 3.00mm Carbide AlTiN
Supplier:  Grainger 16T613
Encompass_Preferred
The Grainger 16T613 is a 3M Scotch-Brite General Purpose Hand Pad that can be used for a variety of cleaning applications. This 8" x 4" x 1/2" hand pad has an open web construction with non-scratching fibers that are gentle on surfaces. It effectively removes grease, grime, food residue, and more from pots, pans, dishes, countertops, appliances, and other surfaces. The pad is durable and long-lasting. It can be used wet or dry. This hand pad from 3M provides versatile cleaning performance.
Catalog No. 50-276-7111