Learn More
Grainger Drill Mill 3.00mm Carbide Bright
Supplier:  Grainger 16T614
Encompass_Preferred
The Grainger 16T614 is a 3/4 HP, 115V, 1725 RPM, 56C Frame, Open Drip Proof, Single Phase, Ball Bearing Electric Motor. It has a NEMA standard 56C frame with a 1.15 service factor. The motor is rated for continuous duty applications with a 40 degree C ambient temperature rating. It has Class F insulation and a C-face endshield for easy mounting.
Catalog No. 50-276-5325