Learn More
Grainger Drill Mill 5.00mm Carbide AlTiN
Supplier:  Grainger 16T627
Encompass_Preferred
The Micro 100 1/4" Shank Threading Tool is a high speed steel threading tool with a 60 degree threading insert. It has a 1/4" shank diameter and is designed for internal threading operations. The tool comes with one DCMT 060204 threading insert which provides a sharp cutting edge and allows for multiple regrinds. The insert is made from micro-grain carbide for wear resistance and strength. This threading tool is suitable for use on most common materials including steel, stainless steel, cast iron, aluminum, brass, bronze, plastics and more. It provides an economical solution for internal threading applications.
Catalog No. 50-276-6806