Learn More
Grainger Drill Mill 5/16 Carbide Bright
Supplier:  Grainger 16T583
Encompass_Preferred
The Micro 100 Drill Mill Carbide is a carbide end mill designed for milling and drilling operations. It has a 1/4 inch shank diameter and is available in multiple lengths from 1/2 inch to 2 inches. The end mill features a TiAlN coating for increased tool life and heat resistance. It has a 135 degree split point tip to aid in drilling and reduce walking. The end mill has 5 flutes and is capable of milling in all directions. It can machine steels, cast iron, aluminum and other non-ferrous metals. The end mill is manufactured in the USA by Micro 100 Tool Corp.
Catalog No. 50-276-6049