Learn More
Medchemexpress LLC Drisapersen sodium | 1181666-20-5 | 98.6% | 7395.0 | C211H256N76Na19O119P19S19 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY145724A5MG
Encompass_Preferred
Drisapersen sodium is an antisense oligonucleotide that induces exon 51 skipping during dystrophin pre-mRNA splicing, enabling synthesis of partially functional dystrophin and supporting research into Duchenne muscular dystrophy. The compound is supplied as the sodium salt and is provided with batch-specific characterization for research use.
  • Antisense oligonucleotide that induces exon 51 skipping.
  • Suitable for Duchenne muscular dystrophy exon-skipping studies.
  • Provided as the sodium salt with molecular formula and molecular weight reported.
  • High purity (98.6%) for experimental consistency.
  • Available in multiple research-scale package sizes.
  • Certificate of analysis provided for batch-specific validation.
Catalog No. 50-002-21383