Learn More
Medchemexpress LLC Dronedarone Hydrochloride | 141625-93-6 | 99.99% | C31H45ClN2O5S | 25 MG
Supplier: Medchemexpress LLC 25MGHY75839
Dronedarone Hydrochloride is a non-iodinated amiodarone derivative that inhibits Na+, K+, and Ca2+ currents. It is intended for research use only and not sold to patients.
- Inhibits Na+, K+, and Ca2+ currents
- Non-iodinated amiodarone derivative
- Potent inhibitor of acetylcholine-activated K+ current
- Blocks IKr and ICa-L
- Reduces incidence of ventricular fibrillation
- Inhibits carotid artery thrombus formation in vivo