Learn More
Medchemexpress LLC Dronedarone (hydrochloride) | 141625-93-6 | 99.99% | C31H45ClN2O5S | 500 MG
SDP
Supplier:  Medchemexpress LLC HY75839500MG
Encompass_Preferred
Dronedarone Hydrochloride is a non-iodinated amiodarone derivative that inhibits Na+, K+, and Ca2+ currents. It serves as a multichannel blocker for atrial fibrillation, demonstrating various inhibitory effects on ion channels and biological activities.
  • Inhibits Na+, K+, and Ca2+ currents
  • Potent inhibitor of acetylcholine-activated K+ current
  • Blocks IKr (IC50=3 μM) and ICa-L (IC50=0.18 μM)
  • Reduces the incidence of ventricular fibrillation in rats
  • Inhibits carotid artery thrombus formation
  • Impairs thrombin- and collagen-induced platelet aggregation
Catalog No. 50-004-33651