Learn More
Medchemexpress LLC Dronedarone impurity 25 hydrochloride | 437651-44-0 | 99.3% | C18H30ClNO3 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHY75572
Encompass_Preferred
Dronedarone impurity 25 hydrochloride is identified as an impurity of Dronedarone. It serves as a drug intermediate.
  • Impurity of Dronedarone
  • Functions as a drug intermediate
  • Off-white to gray solid appearance
  • High purity (99.33%) confirmed by HPLC
Catalog No. 50-003-58482