Learn More
Quantimetrix Dropper Plus, Point of Care Urinalysis Dipstick Control, Set, Level 1&2
SDP
Supplier:  Quantimetrix 144004
Encompass_Preferred
•Liquid, made with human urine•1-month open vial stability at room temperature for maximum portability•18-month shelf life & open vial stability from date of manufacture when stored at 2ºC–8ºCOur Dropper Plus POC Urinalysis Dipstick Control eliminates storage problems and gives you maximum portability by providing one month of room temperature stability – perfect for sites without refrigeration. Easy-to-use dropper bottles make dispensing simple. We include values for all dipstick analytes plus microalbumin and creatinine, and qualitative results for hCG early pregnancy detection test methods. Dropper Plus POC Urinalysis Dipstick Control is designed for use with most urinalysis reagent strips. It can also be used for confirmatory tests and refractometry.Thoughtfully redesigned packaging now includes barcodes on the labels for lot numbers and expiration dates and we’ve made it easier to identify each level.
Catalog No. NC9666606