Learn More
Cayman Chemical DropropIzIn 25g
SDP
Supplier:  Cayman Chemical 3111025 G
Encompass_Preferred
An antitussive agent; reduces the number and intensity of mechanical irritation-induced coughs in conscious cats at 100 and 30 mg/kg, respectively
Catalog No. 50-298-9553