Learn More
Medchemexpress LLC Drotaverine hydrochloride | 985-12-6 | 99.65% | 433.97 | 10 MM 1 ML
SDP
Supplier:  Medchemexpress LLC HY10897410MM/1ML
Encompass_Preferred
Drotaverine hydrochloride is a type 4 cyclic nucleotide phosphodiesterase (PDE4) inhibitor and an L-type voltage-dependent calcium channel (L-VDCC) blocker. It functions by blocking the degradation of 3',5'-cyclic adenosine monophosphate and demonstrates in vivo antispasmodic efficacy without anticholinergic effects. It is intended for research and analytical applications.
  • Acts as a relaxant for pre-contracted airways
  • Exhibits higher selectivity for KCl-induced contractions
  • Inhibits specific binding of nifedipine and diltiazem
  • Improves cognitive impairment and regulates neurotransmitter levels in the brain in animal models
  • Increases brain antioxidant levels and reduces nitrite levels
  • Increases serotonin, dopamine, and norepinephrine levels
Catalog No. 50-003-46122