Learn More
Cayman Chemical DrotaverInhydrochlorIde 100mg
SDP
Supplier:  Cayman Chemical 20944100 MG
Encompass_Preferred
An alkaloid that has been described as an inhibitor of phosphodiesterase 4 and negative allosteric modulator of L-type Ca2+ channels
Catalog No. 50-298-1626