Learn More
TARGETMOL CHEMICALS INC DROXIDOPA 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T254725MG
Encompass_Preferred
Also available in 1 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Droxidopa (L-DOPS) is an orally available prodrug of norepinephrine that is used in the treatment of symptomatic orthostatic hypotension due to neurogenic causes of autonomic failure. purity: 99%
Catalog No. 50-258-0749