Learn More
Selleck Chemical LLC Droxinostat 5mg 99873-43-5 NS 41080
SDP
Supplier:  Selleck Chemical LLC S14225MG
Encompass_Preferred
Droxinostat (NS 41080) is a selective inhibitor of HDAC, mostly for HDACs 6 and 8 with IC50 of 2.47 ?M and 1.46 ?M, greater than 8-fold selective against HDAC3 and no inhibition to HDAC1, 2, 4, 5, 7, 9, and 10. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-831-1