Learn More
AirSea USA 12L UN Combination Packaging, Category A Category B Large Dry Ice Shipper for up to 4L of Biological and Infectious Substances with Inner 12L UN Steel Drum with Lac Lining & Lever Locking Ring – ship both solid and liquid
Supplier:  AirSea USA 424437
Encompass_Preferred
 Biotransporter Cargo - 12L UN Combination Packaging, 4G/Class 6.2 Category A Category B Large Dry Ice Shipper for up to 4L of Biological and Infectious Substances and includes a 12L UN Steel Drum with Lacquer Lining & Lever Locking Ring – approved to ship both solid and liquid hazardous goods substances. UN Marks: 1A2/Y1.8/200 & 1A2/X28/S
Catalog No. NC2101143