Learn More
Alkali Scientific Dry Powder Milk, 1 Kg
SDP
Supplier:  Alkali Scientific DRY002CP
Encompass_Preferred
Dry Powder Milk, 1 Kg, is a non-fat, high-quality milk powder commonly used in laboratory research for microbiological media preparation. It serves as a rich nutrient source, particularly for growing bacteria, fungi, and other microorganisms in controlled environments. The powder is used in a variety of laboratory applications, including fermentation studies, microbiological assays, and the production of standard bacterial culture media. It ensures the reliable growth of microorganisms that require dairy-based nutrients. The 1 kg size is ideal for large-scale media preparation, reducing the frequency of repurchasing smaller packages, and is perfect for facilities with high-demand culture media production.
Catalog No. 50-006-61064