Learn More
Alkali Scientific Dry Powder Milk, 10 Kg
SDP
Supplier:  Alkali Scientific DRY003CP
Encompass_Preferred
Dry Powder Milk, 10 Kg, is a non-fat milk powder designed for use in microbiological and biochemical laboratories for preparing culture media and fermentation processes. This larger bulk size is ideal for high-volume research facilities or production environments that require consistent nutrient-rich media for microbial growth. The powder is an essential ingredient in creating media for the growth of bacteria, fungi, and other microorganisms. It provides the necessary nutrients for metabolic processes and supports the growth of cells in culture. The 10 kg package offers a cost-effective solution for laboratories that require significant quantities of milk powder for routine media preparation and microbial studies.
Catalog No. 50-006-60948