Learn More
Research Products International Corp Dry Powder Milk, 10 KG
SDP
Supplier:  Research Products International Corp M1720010000.0
Encompass_Preferred
Nonfat dry milk for use as a blocking agent during Western Blotting to block non-specific antibody binding and reduce background signal.
Catalog No. 50-223-0242