Learn More
Alkali Scientific Dry Powder Milk, 100 G
SDP
Supplier:  Alkali Scientific DRY001CP
Encompass_Preferred
Dry Powder Milk, 100 G, is a non-fat milk powder used primarily in microbiological culture media and as a nutrient source in laboratory applications. It is commonly employed in the preparation of media for the growth of microorganisms, particularly those requiring a dairy-based nutrient source. The powder is utilized in various applications, including culturing bacteria and fungi, and in fermentation processes. This 100g package size is ideal for small-scale culture media preparation, ensuring precise control over the concentration of nutrients used in experiments. The product is highly versatile and essential in the preparation of high-quality, reproducible media formulations in research laboratories.
Catalog No. 50-006-60359