Learn More
Medchemexpress LLC DS-1040 tosylate | 1335138-89-0 | MFCD31544337 | 99.3% | 465.61 | C23H35N3O5S | 100 MG
SDP
Supplier:  Medchemexpress LLC HY101918100MG
Encompass_Preferred
DS-1040 Tosylate is a research-grade, orally active small-molecule inhibitor of the activated form of thrombin-activatable fibrinolysis inhibitor (TAFIa). It enhances fibrinolysis by inhibiting TAFIa with low-nanomolar potency and is supplied for laboratory research use. Typical applications include in vitro and in vivo studies of fibrinolysis and thromboembolic disease models. Store solid at 4°C; in solvent store at -80°C (6 months) or -20°C (1 month).
  • Orally active small-molecule TAFIa inhibitor.
  • Low-nanomolar potency (human IC50 5.92 nM; rat IC50 8.01 nM).
  • High purity (≈99.3%).
  • Supplied in mg-scale pack sizes for research workflows.
  • Stable under recommended storage conditions.
  • Intended for research use only.
Catalog No. 50-002-46922