Learn More
Medchemexpress LLC DS-1040 tosylate | 1335138-89-0 | MFCD31544337 | 99.3% | 465.61 g/mol | C23H35N3O5S | 50 MG
SDP
Supplier:  Medchemexpress LLC HY10191850MG
Encompass_Preferred
DS-1040 tosylate is an orally active, selective small-molecule inhibitor of activated thrombin-activatable fibrinolysis inhibitor (TAFIa). It enhances fibrinolysis and is used as a research tool to study thromboembolic disease mechanisms. Reported potency includes IC50 values of 5.92 nM for human TAFIa and 8.01 nM for rat TAFIa, and the substance is supplied with supporting analytical documentation.
  • Selective inhibition of activated thrombin-activatable fibrinolysis inhibitor (TAFIa).
  • Potent in vitro activity with low-nanomolar IC50 values for human and rat TAFIa.
  • Provided as a tosylate salt to improve handling and stability.
  • High reported purity suitable for research applications.
  • Accompanied by datasheet, COA, and SDS documentation.
Catalog No. 50-002-46920