Learn More
Takara Bio Living Colors® DsRed Polyclonal Antibody, 100 ul
SDP
Supplier:  Takara Bio 632496
Encompass
The Living Colors DsRed Polyclonal Antibody has been raised against DsRed-Express, a variant of Discosoma sp. red fluorescent protein. This antibody recognizes DsRed-Express, DsRed-Express2, DsRed-Monomer, mCherry, DsRed2, E2-Crimson, tdTomato, mStrawberry, and mBanana, and both N- and C-terminal fusion proteins containing these fluorescent proteins in mammalian cell lysates. 100ul
Catalog No. NC9580775