Learn More
Medchemexpress LLC DS86760016 | 1853176-89-2 | 99.3% | 243.45 | C9H11BClNO4 | 25MG
SDP
Supplier:  Medchemexpress LLC HY12467925MG
Encompass_Preferred
A small-molecule leucyl-tRNA synthetase (LeuRS) inhibitor with activity against multidrug-resistant Gram-negative bacteria. Supplied for research use, it has a molecular weight of 243.45 g/mol, chemical formula C9H11BClNO4, and typical purity ≥99.3%.
  • Potent inhibitor of leucyl-tRNA synthetase.
  • Active against multidrug-resistant Gram-negative bacteria.
  • High chemical purity suitable for analytical and biological studies.
  • Characterized molecular weight of 243.45 g/mol and formula C9H11BClNO4.
  • For research use only; not for human or clinical use.
Catalog No. 50-002-37211