Learn More
Medchemexpress LLC DS86760016 | 1853176-89-2 | 99.3% | 243.45 g/mol | C9H11BClNO4 | 10MG
SDP
Supplier:  Medchemexpress LLC HY12467910MG
Encompass_Preferred
This small-molecule leucyl-tRNA synthetase (LeuRS) inhibitor is intended for antibacterial research and has demonstrated activity against multidrug-resistant Gram-negative pathogens in vitro and in vivo. It is supplied as a high-purity research reagent (CAS 1853176-89-2) with molecular formula C9H11BClNO4 and molecular weight 243.45 g/mol. Solubility and storage recommendations are provided for laboratory use.
  • Potent leucyl-tRNA synthetase inhibitor with demonstrated antibacterial activity.
  • High purity suitable for research applications.
  • Characterized by a defined molecular weight and chemical formula.
  • Soluble in DMSO and water at high concentration with ultrasonic assistance.
  • Recommended sealed, refrigerated storage and low-temperature solvent storage.
  • Available in small-mass quantities for laboratory studies.
Catalog No. 50-002-37188