Learn More
Medchemexpress LLC DSHS00884 | 675104-49-1 | 99.8% | 308.38 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY11379410MM/1ML
Encompass_Preferred
DSHS00884 is a potent human papillomavirus E6 inhibitor with an IC50 of 10 μM. It stabilizes p53 levels in HPV-positive cells by blocking E6-mediated p53 degradation.
  • Stabilizes p53 levels in HPV-positive cells
  • Blocks E6-mediated p53 degradation
Catalog No. 50-004-37512