Learn More
Medchemexpress LLC Dshs00884 (ssya10-001) | 675104-49-1 | 99.8% | 308.38 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY11379450MG
Encompass_Preferred
DSHS00884 is a potent human papillomavirus E6 inhibitor with an IC50 of 10 μM. It stabilizes p53 levels in HPV-positive cells by blocking E6-mediated p53 degradation with an IC50 of 2.5-25 μM. This product is for research use only.
  • Potent human papillomavirus E6 inhibitor
  • Stabilizes p53 levels in HPV-positive cells
  • Blocks E6-mediated p53 degradation
  • For research use only
Catalog No. 50-004-37225