Learn More
Medchemexpress LLC Dshs00884 (SSYA10-001) | 675104-49-1 | 99.8% | 308.38 | C12H12N4O2S2 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1137945MG
Encompass_Preferred
DSHS00884 (SSYA10-001) is a small-molecule inhibitor of the human papillomavirus E6 protein used for in vitro biochemical and cell-based research. It inhibits E6 activity and stabilizes p53 levels in HPV-positive cells, enabling studies of E6-mediated pathways and p53-dependent responses.
  • Inhibits HPV E6 with an IC50 of 10 μM.
  • Blocks E6-mediated p53 degradation in HPV-positive cells.
  • High purity solid suitable for biochemical and cell-based assays.
  • Chemical formula C12H12N4O2S2; molecular weight 308.38.
  • Powder and solvent storage stability provided for long-term research use.
  • Available in small research pack sizes including 5 mg.
Catalog No. 50-002-63403