Learn More
Apexbio Technology LLC DSLET 75644-90-5 1mg
SDP
Supplier:  Apexbio Technology LLC B50671
Encompass_Preferred
DSLET (CAS 75644-90-5) is a selective agonist of the delta ( ) opioid receptor By binding specifically to this receptor subtype DSLET modulates intracellular signaling pathways involved in nociception and affective states Preclinical studies have demonstrated that DSLET elicits significant biological responses in experimental models of pain and stress Owing to its receptor specificity DSLET is widely utilized in basic research to investigate the roles of opioid receptors in pain modulation and emotional regulation supporting the exploration of therapeutic mechanisms underlying analgesia and mood disorders
Catalog No. 50-000-89039