Learn More
AdipoGen DSP-1
SDP
Supplier:  AdipoGen CDXD0391M005
Encompass_Preferred
Chemical. CAS 1609395-29-0. Formula C28H16FNO8. MW 494.44. Synthetic Fluorescent probe for detection of H2S2 and hydrogen polysulfides in aqueous buffers.
Catalog No. 50-256-1094